pH/mV/°C meter, handheld, HQd (HQ11d)
Catalog # HACHHQ11D53000000
供应商: HACH
规格
- Temperature range:−10...+110 °C
- Description:HQ11d portable pH/ORP meter (without electrode)
- Seq:01
- Model:HQ11d
- Temperature accuracy:± 0,3 °C
- Temperature resolution:0,1 °C
- Compliance:IP67, GLP
- Connections:One M12 digital electrode input
- Data logging capacity:500 results
- Display:LCD (240×160 pixels) with backlight
- Outputs:USB
- Power supply:4×AA batteries
- Weight:0,32 kg
- W×D×H:95×36×197 mm
- mV accuracy:±0,1
- mV range:±1500
- mV resolution:0,1
- pH calibration:4-point calibration
- pH range:0 - 14
- pH resolution:0,001 - 0,1 (selectable)
- Pk:1
规格
关于此项目
This portable pH/ORP meter combines reliability, flexibility and ease of use. Interchangeable IntelliCAL™ electrodes are recognised automatically and store all relevant data. Ideal for measuring critical water quality parameters.
- Single input channel
- Outstanding reliability and simple handling
- Reliable results from inaccessible measurement locations and over long distances - even for pH
- Automatic temperature compensation
- Full GLP data management
Kits including electrodes for different applications are available. Rugged, nearly indestructible outdoor versions with multiple cable lengths enable measurements even at previously inaccessible locations.
Delivery: Meter only (without electrode) is supplied with 4 AA batteries, quick-start guide, and user manual. The kits additionally include a pH electrode and reagents.
Additional IntelliCAL™ electrodes are available on request - please contact your local Avantor sales office for further information.
Additional IntelliCAL™ electrodes are available on request - please contact your local Avantor sales office for further information.