PBS buffer
PBS buffer 10X, includes 0,1 M phosphate buffer with 27 mM KCl and 1,37 M NaCl, 1.000 mL
Catalog # GEHEBR-1006-72
Supplier: GE HEALTHCARE
Jump to:
Product Details & Documents
Phosphate buffered saline solution, available with or without detergent, recommended as running buffer for small molecule analysis in Biacore systems. Concentrated stock solution, yielding pH 7,4 when diluted 10X.
- Can be stored at room temperature, precipitation can occur at low temperature
- Supplemented with DMSO
Documents
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Specifications
Specifications
- Pk:1.000 mL
- Description:PBS buffer 10X, includes 0,1 M phosphate buffer with 27 mM KCl and 1,37 M NaCl
- Cat. No.:GEHEBR-1006-72
- For use with:Interaction analyses in Biacore systems except Biacore Flexchip