Order Entry
China
ContactUsLinkComponent
VWR®, Acetate 1000 mg/L in water standard for ion chromatography
VWR®, Acetate 1000 mg/L in water standard for ion chromatography
Catalog # VWRC84951.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Acetate 1000 mg/L in water standard for ion chromatography
Catalog # VWRC84951.180
Supplier:  VWR Chemicals
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Concentration:
    1000 mg/l
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Acetate standard solution
  • Contents:
    CH₃COOH in H₂O

Specifications

About this item

Certifications: Traceable to NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities. Supplied with Certificate of Analysis.