Hydrophobic interaction chromatography, Bio-Beads SM-2
Catalog # BRAD152-3920
Supplier: BIO RAD
CAS Number:
Specifications
- Pk:100 g
- Description:Bio-Beads SM-2 adsorbents, non polar polysterene, 300 - 1,180 µm bead size, for organics < 2000 MW, 100 g
- Environmentally Preferable: