Order Entry
China
ContactUsLinkComponent
Sealing film and foil for PCR plates, Microseal®
  BRADMSB-1001
 :  BIO RAD
Sealing film and foil for PCR plates, Microseal®
  BRADMSB-1001
 :  BIO RAD
  • Optical property:
    Optically clear
  • Description:
    Adhesive seals, Microseal® 'B', optically clear seal, −40...+100 °C
  • Seq:
    02
  • Pk:
    100
Microseal® films and Microseal® seals are used for the 96-well PCR plates and tubes.