Order Entry
China
Orders LinkContactUsLinkComponent
Empty columns, Poly-Prep®
Empty polypropylene columns, Poly-Prep®, 90 mm high, 2 ml bed volume, with end caps and tip closures
  BRAD731-1550
Marketsource
 :   BIO RAD
Empty columns, Poly-Prep®
  BRAD731-1550
 :  BIO RAD
Marketsource
Empty columns, Poly-Prep®
Empty polypropylene columns, Poly-Prep®, 90 mm high, 2 ml bed volume, with end caps and tip closures
1 * 50 件
  • Description:
    Empty polypropylene columns, Poly-Prep®, 90 mm high, 2 ml bed volume, with end caps and tip closures