Order Entry
China
ContactUsLinkComponent
 
HPLC columns, Acclaim™ Trinity™ P2
Catalog # HYPE085434
undefined
HPLC columns, Acclaim™ Trinity™ P2
Catalog # HYPE085434

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Column Format:
    Analytical
  • Ø int.:
    3,00 mm
  • Particle Size:
    3 µm
  • Length:
    100 mm
  • Seq:
    04
  • Pk:
    1

Specifications

About this item

The Acclaim™ Trinity™ P2 is a high-performance, silica-based column specifically designed for separation of charge molecules, including pharmaceutical counter ions by HPLC.

  • Desired selectivity for pharmaceutical counterion screening
  • Retention of ionic and ionisable analytes without using ion-pairing reagents
  • Compatibility with Thermo Scientific™ Dionex™ Corona™ Veo™ Charged Aerosol Detector (CAD) and MS detection methods

Particle type: ultra pure, spherical silica

Max. pressure: 6000 psi

Flow rate range: 0,30 to 0,90 ml/min for 3,0 mm ID formats; 0,15 to 0,45 ml/min for 2,1 mm ID formats

Bonded phase: anion-exchange, cation exchange and HILIC

pH range: 2,0 to 8,0