
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate) ≥98.0% (by titrimetric analysis) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:6578-06-9
:
Formula:C₁₅H₁₅BrClN₂O₄P
MDL Number:MFCD00040636
:433.6214 g/mol
Storage Temperature:Freezer
EINECS:229-506-1
UN:2811
ADR:6.1,III
:GHS06
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Alkaline Phosphatase detection test:to pass test