Order Entry
China
ContactUsLinkComponent
Sodium thiocyanate, Reagent Grade
Irritant
Sodium thiocyanate, Reagent Grade
Catalog # VWRC0861-12KG
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Sodium thiocyanate, Reagent Grade
Catalog # VWRC0861-12KG
Supplier:  VWR Chemicals
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    12 kg
  • Seq:
    4187

Specifications

About this item

 :
 :81.07 g/mol
Merck Index:13,09401
Melting Pt:287 °C
Density:1.735 g/cm³ (20 °C)
EINECS:208-754-4
 :GHS07
Storage Temperature:Ambient
CAS Number:540-72-7
Formula:NaSCN
MDL Number:MFCD00011123