Dithiothreitol (DTT, Cleland's reagent) 1 mol/l in aqueous solution, Sigma-Aldrich®
Catalog # SIAL43816-250ML
Supplier: SIGMAALDRICH
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- Seq:2590
Specifications
About this item
:
:154.25 g/mol
EINECS:222-468-7
CAS Number:3483-12-3
:27565-41-9
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00004877