Order Entry
China
ContactUsLinkComponent
1-Ethoxy-1-(tributylstannyl)ethene 95%
Toxic
Health
Environment
1-Ethoxy-1-(tributylstannyl)ethene 95%
Certificates
undefined
Toxic
Health
Environment
1-Ethoxy-1-(tributylstannyl)ethene 95%

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4750 to 1.4770 (20°C, 589 nm)
  • GC:
    ≥94 %

Specification Test Results

Related Information