Order Entry
China
ContactUsLinkComponent
 
Bathophenanthrolinedisulphonic acid disodium salt hydrate 98%
  ACRO164050010
 :  
undefined
Bathophenanthrolinedisulphonic acid disodium salt hydrate 98%
  ACRO164050010
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to beige to pink Powder
  • Infrared spectrum:
    Conforms
  • Water:
    2 to 6 % (K.F.)
  • UV Lambda max:
    276 to 284 nm (in water)
  • Specific extinction E (1%/1cm):
    ≥710 (at lambda max)
  • Titration with HClO4:
    97.5 to 102.5 % (on dry substance)
  • Chloride (Cl):
    ≤50 ppm

 

 

 :
 :536.48 g/mol (anhy)
 :>300 °C
 :Ambient
 :258-152-0
 :52746-49-3
 :53744-42-6
 :C₂₄H₁₄N₂Na₂O₆S₂·H₂O
 :MFCD00149310