1-Boc-piperidine-4-carboxylic acid
Catalog # CHLCR02213320133
Supplier: LOCAL DEALS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:229.3 g/mol
CAS Number:84358-13-4
Formula:C₁₁H₁₉NO₄
MDL Number:MFCD00076999