
Dextran sulphate sodium salt, M.W. ~ 500,000 50% for biotechnology
Catalog # VWRCE516-100ML
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- APHA (1:10):Tan to yellow haze-free liquid
- DNase:none detected
- RNase:none detected
Specification Test Results
About this item
:
:GHS07
CAS Number:9011-18-1
Formula:(C₆H₇Na₃O₁₄S₃)n
MDL Number:MFCD00081551