Order Entry
China
ContactUsLinkComponent
Sodium hydroxide 0.25 mol/l (0.25 N) in aqueous solution
Irritant
Sodium hydroxide 0.25 mol/l (0.25 N) in aqueous solution
Catalog # HACH1476353
Supplier:  HACH
CAS Number:  
Irritant
Sodium hydroxide 0.25 mol/l (0.25 N) in aqueous solution
Catalog # HACH1476353
Supplier:  HACH
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :40 g/mol
Boiling Pt:∼105 °C (1013 hPa)
Density:1.02 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
 :GHS07
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548