跳转到 页眉
跳转到 主要内容
跳转到 页脚
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium thiosulphate
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution
HACH2700832
:
HACH
:
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution
HACH2700832
:
HACH
:
Pk:
1
Seq:
0001
:
:
158.11 g/mol
:
Ambient
:
231-867-5
:
7772-98-7
:
Na₂S₂O₃
:
MFCD00003499