¥0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
China
ContactUsLinkComponent
Sodium dodecyl sulphate (SDS)
Sodium dodecyl sulphate (SDS) 100 g/l in aqueous solution
Sodium dodecyl sulphate (SDS) 100 g/l in aqueous solution
Sodium dodecyl sulphate (SDS) 100 g/l in aqueous solution
INVT24730020
:
INVITROGEN
:
Sodium dodecyl sulphate (SDS) 100 g/l in aqueous solution
INVT24730020
:
INVITROGEN
:
Pk:
1 L
Seq:
5037
:
:
288.38 g/mol
:
1…1.1 g/cm³ (20 °C)
:
Ambient
:
205-788-1
:
151-21-3
:
CH₃(CH₂)₁₁OSO₃Na
:
MFCD00036175