Order Entry
China
ContactUsLinkComponent
Zinc sulphate 0.05 M ZnSO4 ≥99% volumetric solution, Fluka™
Irritant
Environment
Zinc sulphate 0.05 M ZnSO4 ≥99% volumetric solution, Fluka™
Catalog # HONC34448-1L
Supplier:  HONEYWELL
CAS Number:  
Irritant
Environment
Zinc sulphate 0.05 M ZnSO4 ≥99% volumetric solution, Fluka™
Catalog # HONC34448-1L
Supplier:  HONEYWELL
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002

Specification Test Results

About this item

 :
Boiling Pt:100 °C (solvent)
Melting Pt:<0 °C
Density:1.005 g/cm³
Flash Pt: Does not flash
Storage Temperature:Ambient
EINECS:231-793-3
ADR:9,III
 :287.56 g/mol
 :GHS07
 :GHS09
CAS Number:7446-20-0
Formula:ZnSO₄·7H₂O
MDL Number:MFCD00011302