Methyl-2-amino-3-chlorobenzoate
Catalog # CHLCR00920510134
Supplier: LOCAL DEALS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:185.6100 g/mol
CAS Number:77820-58-7
Formula:C₈H₈ClNO₂
MDL Number:MFCD06797833