DPD chlorine testing systems, SwifTest™
Supplier: HACH
Certificates
About this item
- Fast, easy reagent addition - ideal for high volume testing
- Just squeeze to add powder, no need to open reagent containers
Delivery: Dispenser is supplied with 1 refill vial with enough reagent for approximately 250 tests.