0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Electrodes
ISE Electrodes
Accessories for ion selective electrodes (Ammonia) for HQd Meter
Accessories for ion selective electrodes (Ammonia) for HQd Meter
Accessories for ion selective electrodes (Ammonia) for HQd Meter
Catalog # HACH2980699
Supplier:
HACH
Expand image
Accessories for ion selective electrodes (Ammonia) for HQd Meter
Catalog # HACH2980699
Supplier:
HACH
Specifications
For:
Determining very low ammonium levels
Description:
Ammonium Ionic Strength Adjustor (ISA), 100 powder pillows
Seq:
04
Pk:
1
Specifications
About this item
Product Details
Product Details