Comparator kits for water testing
Supplier: HACH
Certificates
About this item
Qualitative results using traditional colour comparator disc methods.
- Portable - ideal for field testing
- Simple fast method
- Kits contain all required reagents and apparatus in a rugged carrying case
Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.