Specifications
- 说明:Fused silica retention gaps for select mineral oil with internal Ø 0,53 mm and length 2,5 m
- 顺序:09
- 包装规格:5
Specifications
About this item
The fused silica retention gaps has three different polarities, uncoated for universal applicability. Dimensions equal to 0,53 mm int. Ø fused silica provide easy installation.
- Easy to install
- Single continuous length with leak free connections
These fused silica retention gaps with universal CP-Quick-Seal connectors that fit all capillary column sizes. UltiMetal™ retention gaps are complete with coupled high temperature glass-lined connector for ease-of-use.