Order Entry
China
Orders LinkContactUsLinkComponent
Separating funnels, Squibb, PYREX®
Separating funnels, Squibb, PYREX®
500 ml
Catalog # CORN6404-500
Separating funnels, Squibb, PYREX®
Catalog # CORN6404-500
Separating funnels, Squibb, PYREX®
Separating funnels, Squibb, PYREX®
500 ml
Price
Inquire For Price
Log in for availability
1 * 4 件
Product Details & Documents
PYREX® Borosilicate glass.
With PTFE stopcock and linear, high-density, closed-bottom polyethylene stoppers which conform to [ST] stopper dimensions.
Documents
Specifications
  • Capacity:
    500 ml
  • Seq:
    04