Desiccators with 24/29 stopcock, PYREX®
Supplier: CORNING SCIENCE PRODUCTS
Certificates
About this item
These PYREX® desiccators have a rugged 24/29 standard tapered stopcock with a tooled hose connection that serves as the vacuum inlet.
- The tubulation Ext.Ø is approximately 10 mm
- Material is Borosilicate Glass Code 7740
- This design eliminates the need for grease
It is designed so that its manipulation is not affected by the vacuum. To introduce vacuum into the desiccator, loosen the stopcock to disengage the PTFE plug from contact with the glass. A half turn from this position closes the desiccator.
Ordering information: Plate is not included.
Caution: Warning: Do not heat or subject to pressure. Rated to 1-atmosphere vacuum.
Specifications
- Catalog No:
- CORN3121-250
- CORN3121-150
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing