Heat resistant gloves, TempDex 710
Supplier: MAPA
Certificates
About this item
These textured nitrile gloves with seamless textile support provide high dexterity and thermal protection.
- Thin, anatomically shaped glove ensures dexterity
- Excellent abrasion resistance: Longer lasting
- Special nitrile grip coating facilitates work in moderately oily environments
- Dot embossing for better insulation and better grip
- Heat resistant up to a contact temperature of 125 °C
Ideal for handling composite materials after heat processing, handling moulds in the ceramic industry, handling hot steel after welding, as well as for handling hot piping. Also suitable for maintenance of pipes and heating equipment.
Certifications: EN 388:2016 (4111X), EN 407 (X1XXXX); Cat. II
Specifications
- Catalog No:
- MAPA34710027
- MAPA34710021
- Model:
- 710
- 710
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Finish:
- Dot coating on palm and fingers
- Dot coating on palm and fingers
- Cuff style:
- Elastic
- Elastic
- EN norm:
- EN 388 (4111X), EN 407 (X1XXXX)
- EN 388 (4111X), EN 407 (X1XXXX)