Order Entry
China
ContactUsLinkComponent
Bottles, narrow neck, square,with screw cap, PYREX®
Bottles, narrow neck, square,with screw cap, PYREX®
Catalog # CORN1396-500
undefined
Bottles, narrow neck, square,with screw cap, PYREX®
Catalog # CORN1396-500
Specifications
  • Thread:
    GL 45
  • Capacity:
    500 ml
  • Height:
    176 mm
  • Seq:
    05
  • Pk:
    10
Specifications
About this item
Made of PYREX® borosilicate glass, with linerless, one-piece, orange PP screw cap with a drip-free pouring ring.
  • Exceptional chemical and thermal resistance
  • Rounded edges allow easy cleaning
  • White graduations and a large marking spot
  • Glass ridge moulded into the bottle indicates full capacity
  • Autoclavable
Square design saves 13 to 20% of shelf space, and slightly convex walls minimise glass-to-glass contact during autoclaving.
Further PP coloured screw caps for ease of sorting and identification, vented membrane screw caps with 0,22 µm PTFE hydrophobic membrane, PBT solid or open top screw caps and ETFE pouring rings are optionally available.
Caution: Do not tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage. Square bottles should not be used with bottle top filter units, or in other applications involving vacuum pressure as breakage may occur.