Order Entry
China
ContactUsLinkComponent
Automated pipetting system, VIAFLO 96/384
Automated pipetting system, VIAFLO 96/384
Certificates
undefined
Automated pipetting system, VIAFLO 96/384
Specifications
  • Catalog No:
  • INTL6031
  • INTL6001
  • Model:
  • VIAFLO 384
  • VIAFLO 96
  • No. of pipette channels:
  • 96, 384
  • 96
  • Power supply:
  • 100 - 240 V, 50/60 Hz
  • 100 - 240 V, 50/60 Hz
  • W×D×H of system:
  • 420×300×540 mm
  • 420×300×540 mm
  • Weight of system:
  • 27,7 kg
  • 25,7 kg
Specifications
Pipetting heads
Product Information