Order Entry
China
ContactUsLinkComponent
Vacuum gauge set DCP 3000 + VSP 3000
Vacuum gauge set DCP 3000 + VSP 3000
  VACU20683190
   Safer Human and Environmental Health
undefined
Vacuum gauge set DCP 3000 + VSP 3000
  VACU20683190
   Safer Human and Environmental Health

 

  • Accuracy:
    1×103 mbar (7,5×102 Torr)
  • Measuring principle:
    Thermal conductivity (pirani), chemically resistant plastics / ceramics
  • Protection class:
    IP20
  • Description:
    Vacuum gauge, DCP 3000 with VSP 3000 sensor
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Gauge head:
    up to four gauge heads VSP 3000 (Atm. - 10-3 mbar) can be simultaneously connected
  • Housing:
    robust plastic housing with good chemical resistance
  • Max. temperature of media at the gauge head:
    +80 °C
  • Permitted ambient temperature for operation:
    10 − 40 °C
  • Permitted ambient temperature for storage:
    10 − 60 °C
  • Weight:
    0,44 kg
  • W×D×H:
    144×124×114 mm
  • Pk:
    1

 

 

The DCP 3000 is an outstanding vacuum gauge with large illuminated display. The brand new VSP 3000 vacuum sensor is based on thermal conductivity (Pirani) and offers a wide measurement range from atmospheric pressure down to 10 to 3 mbar. The wetted parts are made of chemically resistant plastics (PBT, PUR) and alumina ceramics. It features a much better chemical resistance and ruggedness than conventional Pirani sensors with their fragile metallic spiral-wound filament. Simultaneous connection of up to eight vacuum sensors allows flexible measuring on the spot. The DCP 3000 communicates with external components via the VACUU·BUS® digital bus system. It is characterised by fully automatic configuration, unified plug connections and cable lengths of up to 30 m. In addition the DCP 3000 offers relative pressure measurements (VSK 3000 as reference) and a data logger feature for up to 32000 data values.

  • Up to four gauge heads VSK 3000 (Atm. 0,1 mbar) and four VSP 3000 (Atm. 10 to 3 mbar) can be simultaneously connected
  • Rugged VSP 3000 vacuum sensor made of plastics and ceramics with high chemical resistance
  • Rugged, splash-water proof VSP 3000 vacuum sensor, also for rough operating conditions
  • Wide measurement range from atmospheric pressure to fine vacuum
 : Suitable complete packages for fine vacuum control are available, for more information, please contact Avantor.