
2,6-Dimethylcyclohexanol (mixture of isomers) 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5337-72-4
:
Formula:C₈H₁₆O
MDL Number:MFCD00001502
Density:0.94 g/cm³
Flash Pt:55 °C
Storage Temperature:Ambient
EINECS:226-264-9
UN:1987
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4585 to 1.4615 (20°C, 589 nm)
- GC:≥98.5 %