1,4-Dioxaspiro[4.5]decan-8-one
Catalog # CHLCR00315610121
Supplier: LOCAL DEALS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:156.18 g/mol
CAS Number:4746-97-8
Formula:C₈H₁₂O₃
MDL Number:MFCD00010214