Order Entry
China
ContactUsLinkComponent
1,4-Dioxaspiro[4.5]decan-8-one 98%
 
1,4-Dioxaspiro[4.5]decan-8-one 98%
  ACRO235640250
 :  
undefined
 
1,4-Dioxaspiro[4.5]decan-8-one 98%
  ACRO235640250
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to beige Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    70 to 74 °C
  • GC:
    ≥97.5 %

 

 

 :
 :156.18 g/mol
 :74…76 °C
 :GHS07
 :Ambient
 :4746-97-8
 :C₈H₁₂O₃
 :MFCD00010214