Order Entry
China
ContactUsLinkComponent
HPLC columns, Hypersil GOLD™ Amino
HPLC columns, Hypersil GOLD™ Amino
Catalog # HYPE25705-254630
undefined
HPLC columns, Hypersil GOLD™ Amino
Catalog # HYPE25705-254630

Specifications

  • Ø int.:
    4,60 mm
  • Particle Size:
    5 µm
  • Pore size:
    175 Å
  • Length:
    250 mm
  • Seq:
    35
  • Phase:
    Amino
  • Pk:
    1

Specifications

About this item

A high performance aminopropyl phase that gives excellent chromatographic properties in three modes: Weak anion exchange, reversed phase and normal phase.

  • Retains anions and organic acids in weak anion exchange mode when used with common buffers and an organic modifier
  • Excels for carbohydrate analysis in reversed phase mode
  • Offers an alternative selectivity to silica columns in normal phase chromatography