2-Propanol
25 L
目录编号 CHLC431091
供应商: LOCAL DEALS
Jump to:
Product Details & Documents
同义词: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
- CAS号:67-63-0
- 化学式:(CH₃)₂CHOH
- MDL 编号:MFCD00011674
- Molecular Weight:60.1 g/mol
- 默克索引:13,05228
- 沸点:82 °C (1013 hPa)
- 熔点:–89 °C
- 密度:0.786 g/cm³ (20 °C)
- 闪点:12 °C
- 储存温度:Ambient
- EINECS:200-661-7
- UN:1219
- ADR:3,II
- REACH:01-2119457558-25
规格
规格
- Pk:25 L
- Cat. No.:CHLC431091
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Safer Human and Environmental Health
规格
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



