Order Entry
China
ContactUsLinkComponent
 
D-(+)-Glucose 40% (w/v) in aqueous solution, Ultra Pure Grade for biochemistry
Catalog # VWRCE701-100ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
D-(+)-Glucose 40% (w/v) in aqueous solution, Ultra Pure Grade for biochemistry
Catalog # VWRCE701-100ML
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear to slightly yellow liquid
  • Glucose:
    35 – 40%

Specification Test Results

About this item

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose
 :
 :180.16 g/mol
Storage Temperature:Ambient
EINECS:200-075-1
CAS Number:50-99-7
 :492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912