Order Entry
China
ContactUsLinkComponent
Supelco® Organic Reference Standard, Methyl Arachidate
Supelco® Organic Reference Standard, Methyl Arachidate
Catalog # SUPL10941-1G
Supplier:  SUPELCO
CAS Number:  
undefined
Supelco® Organic Reference Standard, Methyl Arachidate
Catalog # SUPL10941-1G
Supplier:  SUPELCO
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Standards:
    Analytical standard
  • CAS No.:
    1120-28-1
  • Description:
    Methyl arachidate
  • Seq:
    01
  • Storage conditions:
    –20 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00009014
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Arachidic Acid Methyl Ester; Eicosanoic acid, methyl ester; Icosanoic Acid Methyl Ester; Methyl icosanoate; Methyl icosanoate; Arachidic acid methyl ester; Methyl eicosanoate
  • Melting point:
    45...48 °C (lit.); 46...49 °C
  • Formula:
    CH₃(CH₂)₁₈COOCH₃
  • Molecular weight:
    326,56
  • Flash point:
    Not applicable
  • Boiling point:
    215...216 °C/10 mm Hg (lit.)

Specifications

About this item