Order Entry
China
ContactUsLinkComponent
Benzyldimethyltetradecylammonium chloride hydrate ≥98.0% (by HPLC, titration analysis)
Corrosive
Toxic
Environment
Benzyldimethyltetradecylammonium chloride hydrate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA5160-5G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
Toxic
Environment
Benzyldimethyltetradecylammonium chloride hydrate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA5160-5G
Supplier:  TCI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (TPBNa Method):
    min. 98.0 % (calcd.on anh.substance)
  • Solubility in Water:
    almost transparency
  • Water:
    max. 12.0 %

Specification Test Results

About this item

 :
 :368.05 g/mol (anhy)
Merck Index:14,01059
EINECS:205-352-0
UN:2923
ADR:8,III
 :GHS05
 :GHS06
 :GHS09
CAS Number:139-08-2
Formula:C₂₃H₄₂ClN·H₂O
MDL Number:MFCD00011771