Order Entry
China
ContactUsLinkComponent
 
HPLC columns, Dionex™ OmniPac™
Catalog # HYPE042191
HPLC columns, Dionex™ OmniPac™
Catalog # HYPE042191

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Column Format:
    Analytical
  • Ø int.:
    4,00 mm
  • Particle Size:
    8,5 µm
  • Description:
    PCX-500
  • Length:
    250 mm
  • Seq:
    07
  • Pk:
    1

Specifications

About this item

OmniPac is a range of latex-based columns. Ideal for the seperation of the high molecular weight organic acids.

  • Delivers optimal separation of very hydrophobic and halogenated anions
  • Provides simultaneous separation of neutral and ionic species
  • Unique selectivity for polar and ionic organic analytes

This polymer-based mixed-mode column is solvent compatible and stable over the entire pH range of 1 to 14. Solvents may be used to affect selectivity as well as to remove contaminants to extend column lifetime. The PAX-500 and PCX-500 columns separate analytes through both ion exchange and reversed-phase mechanisms, due to their higher reversedphase capacity relative to the PAX-100 and PCX-100 columns.