


Ammonium peroxodisulphate (APS) ≥98.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
:7727-54-0
:
:(NH₄)₂S₂O₈
:MFCD00003390
:228.2 g/mol
:13,00545
:120 °C
:1.98 g/cm³ (20 °C)
:231-786-5
:1444
:5.1,III
:GHS03
:GHS08
:GHS07
:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Meets ACS Reagent Chemical Requirements:≥98.0%
- For Laboratory,Research,or Manufacturing Use:≤0.005%
- Store below 20°C.:≤0.050%
- Assay ((NH₄)₂S₂O₈):≥ 98.0 %
- Insoluble Matter:≤ 0.005 %
- Residue after Ignition:≤ 0.050 %
- Titrable Free Acid (meq/g):≤ 0.04
- Chloride and Chlorate (as Cl):≤ 0.001 %
- Heavy Metals (as Pb):≤ 0.005 %
- Trace Impurities - Iron (Fe):≤ 0.001 %
- Trace Impurities - Manganese (Mn):≤ 0.500 ppm