TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride)
Catalog # PROMH5123
Supplier: PROMEGA
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:157.6 g/mol
Merck Index:12,09902
Melting Pt:150…152 °C
Storage Temperature:Ambient
EINECS:214-684-5
CAS Number:1185-53-1
Formula:NH₂C(CH₂OH)₃·HCl
MDL Number:MFCD00012590