Trypan blue
Glass bottle, 100 mL
Supplier: INVITROGEN
Jump to:
Product Details & Documents
Synonyms: Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, AzidinBlue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)
- CAS Number:72-57-1
- Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
- MDL Number:MFCD00003969
- Molecular Weight:960.82 g/mol
- EINECS:200-786-7
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle
- Pk:100 mL



