跳转到 页眉
跳转到 主要内容
跳转到 页脚
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Boc-4-(hydroxymethyl)piperidine
1-Boc-4-(hydroxymethyl)piperidine
1-Boc-4-(hydroxymethyl)piperidine
1-Boc-4-(hydroxymethyl)piperidine
CHLCR01662010131
:
LOCAL DEALS
:
1-Boc-4-(hydroxymethyl)piperidine
CHLCR01662010131
:
LOCAL DEALS
:
Pack type:
Glass bottle for solids
Pk:
10 g
Seq:
3236
:
:
215.29 g/mol
:
123855-51-6
:
C₁₁H₂₁NO₃
:
MFCD02094488