Buffer, TRIS solution pH 8 (1 mol/L)
TRIS buffer solution pH 8 (1 mol/l), Glass bottle, 1.000 mL
Supplier: INVITROGEN
Buffer, TRIS solution pH 8 (1 mol/L)
Catalog # INVT15568-025
Supplier: INVITROGEN
Marketsource
CAS Number:
Jump to:
Product Details & Documents
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle
- Pk:1.000 mL
- Description:TRIS buffer solution pH 8 (1 mol/l)



