Buffers, sodium phosphate, 0,2 M solutions
Catalog # ALFAJ62733.K2
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- 浓度:0,2 mol/l
- 包装类型:Glass bottle
- 包装规格:1 L
- 说明:Sodium phosphate buffer solution
- 顺序:12
- MDL No.:MFCD00146206
- 自然形状:Liquid
- pH 值:8,0
Specifications
About this item
Physical form: liquid
MDL No.: MFCD00146206