Order Entry
China
ContactUsLinkComponent
Buffers, citrate, 0,5 M solutions
Buffers, citrate, 0,5 M solutions
Catalog # ALFAJ60024.AP
CAS Number:  
Buffers, citrate, 0,5 M solutions
Catalog # ALFAJ60024.AP
CAS Number:  

Specifications

  • Concentration:
    0,5 M
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • CAS No.:
    6132-04-3
  • Description:
    Citrate, buffer solution
  • Seq:
    11
  • MDL No.:
    MFCD00150031
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • Odour/scent:
    Odourless
  • Appearance:
    Colourless
  • pH value:
    4,5

Specifications

About this item

CAS No.: 6132-04-3
Storage temperature: 2 to 8 °C
Appearance: colourless
Odour: odourless
MDL No.: MFCD00150031