¥0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
China
ContactUsLinkComponent
Buffers
Biological Buffers
Buffer, HEPES solution (1 mol/L), Sigma-Aldrich®
Buffer, HEPES solution (1 mol/L), Sigma-Aldrich®
SIAL83264-100ML-F
:
SIGMAALDRICH
:
Buffer, HEPES solution (1 mol/L), Sigma-Aldrich®
SIAL83264-100ML-F
:
SIGMAALDRICH
:
Pack type:
Glass bottle
Pk:
100 mL
Description:
HEPES buffer solution (1 mol/l)