Order Entry
China
Orders LinkContactUsLinkComponent
 

Copper(I) bromide dimethyl sulphide complex 99%

Glass bottle, 10 g
  ACRO213180100
Copper(I) bromide dimethyl sulphide complex 99%
  ACRO213180100
 :  54678-23-8
Copper(I) bromide dimethyl sulphide complex 99%
Copper(I) bromide dimethyl sulphide complex 99%

Glass bottle, 10 g

 :  54678-23-8
 
 
   
1 * 10 克
 
  •  :54678-23-8
  •  :C₂H₆BrCuS
  •  :MFCD00043295
  •  :205.58 g/mol
  •  :132 °C (dec.)
  •  :Ambient
  •  :259-292-5

 

 

 

  • Appearance:
    Off-white to beige to green to grey Crystalline powder and/or chunks
  • Infrared spectrum:
    Conforms
  • Titration Iodometric:
    ≥98.5 %

 

  • Pack type:
    Glass bottle
  • Pk:
    10 g