Supelco® Organic Reference Standard, Glycine hydrochloride solution
Catalog # SUPL55097-5ML-F
Supplier: SUPELCO
CAS Number:
Specifications
- Concentration:100 mM amino acid in 0,1 M HCl
- Pk:5 mL
- Standards:Analytical standard
- CAS No.:6000-43-7
- Description:Glycine hydrochloride solution
- Storage conditions:2…8 °C
- Hazard class description:H290 - H315 - H319
- Shipping conditions:STD
- MDL No.:MFCD00012872
- M-Clarity quality level:MQ100
- Molecular weight:111,53
Specifications
About this item
- GHS signal: GHS05
Caution: P234 - P264 - P280 - P302 + P352 - P305 + P351 + P338 - P332 + P313