DNA (Deoxyribonucleic acid) (from calf thymus), activated
Catalog # GEHE27-4575-01
Supplier: GE HEALTHCARE
CAS Number:
Specifications
- Pk:1
- Seq:6030
- Pk Info:25 units/ml
Specifications
About this item
:
CAS Number:9007-49-2
:73049-39-5
MDL Number:MFCD00130922