Order Entry
China
ContactUsLinkComponent
Disposable columns prepacked with Sepharose™, GST bulk kit
Disposable columns prepacked with Sepharose™, GST bulk kit
Catalog # GEHE27-4570-01
Supplier:  GE HEALTHCARE
undefined
Disposable columns prepacked with Sepharose™, GST bulk kit
Catalog # GEHE27-4570-01
Supplier:  GE HEALTHCARE
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Sepharose™ bulk GST purification kit
  • Pk:
    1 KIT
Specifications
About this item
GST bulk kit is designed for small-scale manual purification of GST-tagged proteins using either column chromatography or batch method. Facilitates optimization studies of expression, solubility conditions and purification parameters of GST-tagged proteins.
  • For rapid purification of GST-tagged proteins using gravity flow columns or using batch purification with high specificity
  • Preserves protein antigenicity and function
  • Yields up to 50 mg of pure protein
GST-tagged proteins can be recovered under mild elution conditions (10 mM glutathione), which preserves functionality of the proteins. The binding capacity of Glutathione Sepharose 4B, which can be greater than 10 mg protein/ml medium, varies with the nature of the tagged protein and the exact binding conditions used.
GST Bulk Kit is designed for small-scale manual purification of GST-tagged proteins using either column chromatography or a batch method. The batch method is more flexible, as purification can be performed with between 10 μl and 2 ml of Glutathione Sepharose 4B.
GST Bulk Kit also facilitates optimization studies of expression, solubility conditions, and purification parameters of GST-tagged proteins.
Ordering information: Includes 10 ml bulk pack of Glutathione Sepharose 4B medium, 5× gravity-flow columns and IPTG GST buffer kit. The kit contains medium and reagents for purification of GST-tagged proteins from milliliter volumes of sample to larger volumes of sample containing as much as 50 mg of GST-tagged protein.